COMH4211B2A2 Combi-Blocks, Inc.

4-(Cyclohexylsulfanyl)butanoic acid

  • MDL number MFCD09259503
  • Cas Number : 1927-54-4
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4451
Availability details for
Quantity
  • Availability
  • Add to Cart