COMH4211B2A6 Combi-Blocks, Inc.

N-(4-Methoxybenzyl)-o-methylhydroxylamine

  • MDL number MFCD18825398
  • Cas Number : 543731-16-4
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4457
Availability details for
Quantity
  • Availability
  • Add to Cart