COMH4211B2AA Combi-Blocks, Inc.

2-(4-Methoxy-pyrazol-1-yl)-phenylamine

  • MDL number MFCD31579539
  • Cas Number : 1854937-26-0
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4469

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4469
Availability details for
Quantity
  • Availability
  • Add to Cart