COMH4211B2AB Combi-Blocks, Inc.

Cyclopropyl-(4-methoxy-3-nitro-benzyl)-amine

  • MDL number MFCD11119683
  • Cas Number : 1042511-70-5
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4470
Availability details for
Quantity
  • Availability
  • Add to Cart