COMH4211B2AC Combi-Blocks, Inc.

Toluene-4-sulfonic acid 2-(2-[2-(tetrahydro-pyran-2-yloxy)-ethoxy]-ethoxy)-ethyl ester

  • MDL number MFCD31381619
  • Cas Number : 69502-33-6
  • Molecular Weight : 388.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4472
Availability details for
Quantity
  • Availability
  • Add to Cart