COMH4211B2AE Combi-Blocks, Inc.

tert-Butyl 2-fluoro-6-iodobenzoate

  • MDL number MFCD27944906
  • Cas Number : 146014-71-3
  • Molecular Weight : 322.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4474
Availability details for
Quantity
  • Availability
  • Add to Cart