COMH4211B2AF Combi-Blocks, Inc.

2-Thiophenecarboxaldehyde, 4-(4-chlorophenyl)-

  • MDL number MFCD06802076
  • Cas Number : 893736-71-5
  • Molecular Weight : 222.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4479
Availability details for
Quantity
  • Availability
  • Add to Cart