COMH4211B2B1 Combi-Blocks, Inc.

4-[2-(3-Methylpiperidin-1-yl)ethyl]aniline

  • MDL number MFCD09884757
  • Cas Number : 1017212-41-7
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4483
Availability details for
Quantity
  • Availability
  • Add to Cart