COMH4211B2B3 Combi-Blocks, Inc.

4-Methyl-2-[(4-methylphenyl)sulfonylamino]-n-(1-phenylethyl)pentanamide

  • MDL number MFCD12318559
  • Cas Number : 1214160-37-8
  • Molecular Weight : 388.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4485
Availability details for
Quantity
  • Availability
  • Add to Cart