COMH4211B2B4 Combi-Blocks, Inc.

2-Chloro-n-(2,2,2-trifluoroethyl)pyridine-3-carboxamide

  • MDL number MFCD09931841
  • Cas Number : 1016512-43-8
  • Molecular Weight : 238.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4490
Availability details for
Quantity
  • Availability
  • Add to Cart