COMH4211B2B6 Combi-Blocks, Inc.

3-Fluoro-4-(4-methoxy-benzyloxy)-benzoic acid

  • MDL number MFCD13811008
  • Cas Number : 1021144-65-9
  • Molecular Weight : 276.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-((4-methoxybenzyl)oxy)benzoic acid
Inchi Code 1S/C15H13FO4/c1-19-12-5-2-10(3-6-12)9-20-14-7-4-11(15(17)18)8-13(14)16/h2-8H,9H2,1H3,(H,17,18)
Inchi Key GFXHDVRTXLFDNH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4493
Availability details for
Quantity
  • Availability
  • Add to Cart