COMH4211B2B8 Combi-Blocks, Inc.

6-Cyclopropylmethoxy-3,4-dihydro-2h-naphthalen-1-one

  • MDL number MFCD16117600
  • Cas Number : 1247447-23-9
  • Molecular Weight : 216.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4499
Availability details for
Quantity
  • Availability
  • Add to Cart