COMH4211B2B9 Combi-Blocks, Inc.

4-Bromo-2-fluoro-benzoic acid benzyl ester

  • MDL number MFCD28406116
  • Cas Number : 437764-46-0
  • Molecular Weight : 309.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 4-bromo-2-fluorobenzoate
Inchi Code 1S/C14H10BrFO2/c15-11-6-7-12(13(16)8-11)14(17)18-9-10-4-2-1-3-5-10/h1-8H,9H2
Inchi Key JTXNSDOLYAYGAB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4502
Availability details for
Quantity
  • Availability
  • Add to Cart