COMH4211B2BD Combi-Blocks, Inc.

Cyclopropyl-(5-methoxy-pyridin-3-ylmethyl)-amine

  • MDL number MFCD24259567
  • Cas Number : 1545091-51-7
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4509
Availability details for
Quantity
  • Availability
  • Add to Cart