COMH4211B2BE Combi-Blocks, Inc.

Benzyl chloromethyl pentanedioate

  • MDL number MFCD31583275
  • Cas Number : 87343-61-1
  • Molecular Weight : 270.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4511
Availability details for
Quantity
  • Availability
  • Add to Cart