COMH4211B2C0 Combi-Blocks, Inc.

1H-Pyrazole, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[[2-(trifluoromethoxy)phenyl]methyl]-

  • MDL number MFCD28718191
  • Cas Number : 1604036-79-4
  • Molecular Weight : 368.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4517
Availability details for
Quantity
  • Availability
  • Add to Cart