COMH4211B2C1 Combi-Blocks, Inc.

6-(2-Isopropoxyethoxy)nicotinic acid

  • MDL number MFCD09808625
  • Cas Number : 1016493-51-8
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4521
Availability details for
Quantity
  • Availability
  • Add to Cart