COMH4211B2C3 Combi-Blocks, Inc.

6-(2-Methoxyphenyl)-1-oxa-6-azaspiro[2.5]octane

  • MDL number MFCD27937142
  • Cas Number : 1823271-74-4
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4523
Availability details for
Quantity
  • Availability
  • Add to Cart