COMH4211B2C4 Combi-Blocks, Inc.

Benzyl (8-aminooctyl)carbamate

  • MDL number MFCD02094514
  • Cas Number : 66095-19-0
  • Molecular Weight : 278.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl (8-aminooctyl)carbamate
Inchi Code 1S/C16H26N2O2/c17-12-8-3-1-2-4-9-13-18-16(19)20-14-15-10-6-5-7-11-15/h5-7,10-11H,1-4,8-9,12-14,17H2,(H,18,19)
Inchi Key XUPJSKSDZXQOJN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4524
Availability details for
Quantity
  • Availability
  • Add to Cart