COMH4211B2C5 Combi-Blocks, Inc.

6-Isobutoxy-3,4-dihydro-2h-naphthalen-1-one

  • MDL number MFCD16155587
  • Cas Number : 221651-66-7
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4525

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4525
Availability details for
Quantity
  • Availability
  • Add to Cart