COMH4211B2C7 Combi-Blocks, Inc.

6-Chloro-9-ethyl-9h-purine

  • MDL number MFCD07440136
  • Cas Number : 5462-86-2
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4529
Availability details for
Quantity
  • Availability
  • Add to Cart