COMH4211B2C8 Combi-Blocks, Inc.

2-(Benzyloxy)-5-bromo-3-chloropyridine

  • MDL number MFCD29113547
  • Cas Number : 1289121-39-6
  • Molecular Weight : 298.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4530
Availability details for
Quantity
  • Availability
  • Add to Cart