COMH4211B2CB Combi-Blocks, Inc.

1-[4-Nitro-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-piperidine

  • MDL number MFCD22573169
  • Cas Number : 1355193-23-5
  • Molecular Weight : 332.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4538
Availability details for
Quantity
  • Availability
  • Add to Cart