COMH4211B2CD Combi-Blocks, Inc.

1-Bromo-2-fluoro-4-(2,2,2-trifluoroethoxy)-benzene

  • MDL number MFCD21142839
  • Cas Number : 1036724-63-6
  • Molecular Weight : 273.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4542
Availability details for
Quantity
  • Availability
  • Add to Cart