COMH4211B2CE Combi-Blocks, Inc.

4-(3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazol-1-yl)propyl)morpholine

  • MDL number MFCD21337775
  • Cas Number : 1092500-87-2
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4549

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4549
Availability details for
Quantity
  • Availability
  • Add to Cart