COMH4211B2CF Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)-5-hydroxybenzamide

  • MDL number MFCD16999337
  • Cas Number : 1243451-03-7
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4553
Availability details for
Quantity
  • Availability
  • Add to Cart