COMH4211B2D2 Combi-Blocks, Inc.

4-((5-Bromo-1h-pyrazol-1-yl)methyl)benzoic acid

  • MDL number MFCD31583013
  • Cas Number : 1616099-11-6
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4560
Availability details for
Quantity
  • Availability
  • Add to Cart