COMH4211B2EB Combi-Blocks, Inc.

4-Bromo-3-fluoro-n-propyl-benzenesulfonamide

  • MDL number MFCD31539561
  • Cas Number : 1055995-82-8
  • Molecular Weight : 296.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-3-fluoro-N-propylbenzenesulfonamide
Inchi Code 1S/C9H11BrFNO2S/c1-2-5-12-15(13,14)7-3-4-8(10)9(11)6-7/h3-4,6,12H,2,5H2,1H3
Inchi Key MPUCFWMCCARHHY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4564
Availability details for
Quantity
  • Availability
  • Add to Cart