COMH4211B2EC Combi-Blocks, Inc.

Methyl 4-(5-bromo-3-nitropyridin-2-yl)benzoate

  • MDL number MFCD31613880
  • Cas Number : 1800345-13-4
  • Molecular Weight : 337.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4565

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4565
Availability details for
Quantity
  • Availability
  • Add to Cart