COMH4211B2ED Combi-Blocks, Inc.

1-(2,2-Difluoropropoxy)-4-iodobenzene

  • MDL number MFCD30616481
  • Cas Number : 2206607-34-1
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4570
Availability details for
Quantity
  • Availability
  • Add to Cart