COMH4211B2EE Combi-Blocks, Inc.

2-Amino-n-ethyl-5-fluorobenzamide

  • MDL number MFCD11132596
  • Cas Number : 1094252-27-3
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4571
Availability details for
Quantity
  • Availability
  • Add to Cart