COMH4211B2EF Combi-Blocks, Inc.

4-Amino-2-(2,2,2-trifluoroethoxy)toluene

  • MDL number MFCD12190448
  • Cas Number : 220996-42-9
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4573

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4573
Availability details for
Quantity
  • Availability
  • Add to Cart