COMH4211B2F1 Combi-Blocks, Inc.

3-(Difluoromethyl)-2-methylaniline

  • MDL number MFCD18389560
  • Cas Number : 61708-25-6
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4576
Availability details for
Quantity
  • Availability
  • Add to Cart