COMH4211B2F2 Combi-Blocks, Inc.

(S)-1-(1-Phenylethyl)urea

  • MDL number MFCD18642763
  • Cas Number : 25144-64-3
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4577
Availability details for
Quantity
  • Availability
  • Add to Cart