COMH4211B2F3 Combi-Blocks, Inc.

2-(2-(2,2,2-Trifluoroethoxy)phenoxy)ethanamine

  • MDL number MFCD17169996
  • Cas Number : 751476-91-2
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4578
Availability details for
Quantity
  • Availability
  • Add to Cart