COMH4211B2F6 Combi-Blocks, Inc.

1-(4-Chloro-2-fluoro-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31539565
  • Cas Number : 2206608-00-4
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4590
Availability details for
Quantity
  • Availability
  • Add to Cart