COMH4211B2F7 Combi-Blocks, Inc.

1-Cbz-4-allylpiperidine

  • MDL number MFCD24856525
  • Cas Number : 1352733-74-4
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4593
Availability details for
Quantity
  • Availability
  • Add to Cart