COMH4211B2F8 Combi-Blocks, Inc.

Cyclopropyl-(4-methoxy-2-methyl-benzyl)-amine

  • MDL number MFCD21187720
  • Cas Number : 1085524-30-6
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4597

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4597
Availability details for
Quantity
  • Availability
  • Add to Cart