COMH4211B2FA Combi-Blocks, Inc.

2-Methyl-3-(tetrahydro-pyran-4-yl)-propionic acid methyl ester

  • MDL number MFCD31583076
  • Cas Number : 2270912-65-5
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4604
Availability details for
Quantity
  • Availability
  • Add to Cart