COMH4211B2FB Combi-Blocks, Inc.

4-Bromo-3-ethoxy-n-(2-hydroxy-ethyl)-benzamide

  • MDL number MFCD30830151
  • Cas Number : 1309682-47-0
  • Molecular Weight : 288.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4606
Availability details for
Quantity
  • Availability
  • Add to Cart