COMH4211B2FC Combi-Blocks, Inc.

3-Fluoro-2-hydroxy-5-nitrobenzaldehyde

  • MDL number MFCD11847517
  • Cas Number : 2923-99-1
  • Molecular Weight : 185.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4608

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4608
Availability details for
Quantity
  • Availability
  • Add to Cart