COMH4211B301 Combi-Blocks, Inc.

5-Bromo-1,3-difluoro-2-(2-methoxy-ethoxy)-benzene

  • MDL number MFCD31613885
  • Cas Number : 1416360-39-8
  • Molecular Weight : 267.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4616
Availability details for
Quantity
  • Availability
  • Add to Cart