COMH4211B302 Combi-Blocks, Inc.

Benzenemethanamine, 3,4-dichloro-n-(phenylmethoxy)-, hydrochloride

  • MDL number MFCD31561568
  • Cas Number : 543730-63-8
  • Molecular Weight : 318.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4618
Availability details for
Quantity
  • Availability
  • Add to Cart