COMH4211B304 Combi-Blocks, Inc.

Methyl 2-(3,5-bis(hydroxymethyl)phenoxy)acetate

  • MDL number MFCD31614063
  • Cas Number : 1107630-95-4
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4621

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4621
Availability details for
Quantity
  • Availability
  • Add to Cart