COMH4211B306 Combi-Blocks, Inc.

(2-Ethyl-5-iodophenyl)methanol

  • MDL number MFCD28969726
  • Cas Number : 1799974-93-8
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4624

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4624
Availability details for
Quantity
  • Availability
  • Add to Cart