COMH4211B309 Combi-Blocks, Inc.

(1-(4-Chlorophenyl)cyclobutyl)methanol

  • MDL number MFCD22571446
  • Cas Number : 50921-41-0
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-(4-chlorophenyl)cyclobutyl)methanol
Inchi Code 1S/C11H13ClO/c12-10-4-2-9(3-5-10)11(8-13)6-1-7-11/h2-5,13H,1,6-8H2
Inchi Key TYJYYNPUIKWSOP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4633
Availability details for
Quantity
  • Availability
  • Add to Cart