COMH4211B30A Combi-Blocks, Inc.

Bromuron

  • MDL number MFCD00017800
  • Cas Number : 3408-97-7
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4634
Availability details for
Quantity
  • Availability
  • Add to Cart