COMH4211B30B Combi-Blocks, Inc.

4-Bromo-n-ethyl-2-methoxy-6-nitroaniline

  • MDL number MFCD31579425
  • Cas Number : 1416554-52-3
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4635
Availability details for
Quantity
  • Availability
  • Add to Cart