COMH4211B30D Combi-Blocks, Inc.

(R)-3-Amino-piperidine-1-carboxylic acid amide hydrochloride

  • MDL number MFCD31583366
  • Cas Number : 2270918-21-1
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4639
Availability details for
Quantity
  • Availability
  • Add to Cart