COMH4211B311 Combi-Blocks, Inc.

Pentanoic acid, 5-[[6-[[(1,1-dimethylethoxy)carbonyl]amino]hexyl]amino]-5-oxo-

  • MDL number MFCD31381682
  • Cas Number : 1456806-42-0
  • Molecular Weight : 330.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4645
Availability details for
Quantity
  • Availability
  • Add to Cart